FMOC-Lys(Cy5)-OH for biomolecular labeling

Supplier: AAT BIOQUEST INC
Warning

5055
76481-746EA 815.92 USD
76481-746
FMOC-Lys(Cy5)-OH for biomolecular labeling
FMOC-Lys(Cy5)-OH

FMOC-Lys(Cy5)-OH is a building block for in-sequence Lys labeling by Cy5.


  • FRET building block
  • Multiple applications

MW: 1121,24 g/mol
CAS Number: 312961-84-5
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/74/35/37377435.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR