FMOC-Lys(5/6-FAM)-OH for biomolecular labeling
Supplier: AAT BIOQUEST INC
Warning
5042
76481-734EA 529.69 USD
76481-734
FMOC-Lys(5/6-FAM)-OH for biomolecular labeling
FMOC-Lys(5/6-FAM)-OH
FMOC-Lys(5/6-FAM)-OH is a building block for in-sequence Lys labeling by FAM. FAM is one of the most common donor dyes for preparing FRET peptides (often paired with TAMRA or TQ2).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/74/29/37377429.jpg