Wide-Mouth Containers, Caplugs
Supplier: Caplugs
240-9750-W3L 240-9400-G3L 240-9750-G3L 240-9500-W3L 240-9250-G3L 240-9400-W3L 240-9950-G3L 240-9250-W3L 240-9950-W3L 240-9500-G3L
76520-694CT 74.22 USD
76520-694 76520-696 76520-710 76520-698 76520-700 76520-702 76520-704 76520-706 76520-708 76520-692
Wide-Mouth Containers, Caplugs
Bottles Plastic Bottles
Wide-Mouth Containers with Screw Caps are designed for storing biological fluids and pathology specimens. These containers are also useful for almost any miscellaneous storage needs, such as at laboratory or nursing stations.
  • Precision-molded from polypropylene
  • Containers can withstand boiling and freezing
Containers are chemically resistant to 95% ethyl alcohol and 10% formalin, and some other commonly used organic solvents such as kerosene and isopropyl alcohol. The screwcaps are lined with XPE foam to help minimize evaporation during long-term storage and to help eliminate leakage during shipping. 