VWR® Plus Flammable Material Storage Freezers
Supplier: VWR
VWR®
FFS-30
10819-706EA 11909.1 USD
10819-706
VWR® Plus Flammable Material Storage Freezers
Freezers Flammable Material Freezers
The VWR® Plus Series Flammable Storage Freezers are designed to meet NFPA and OSHA guidelines 45 and 70 for the refrigerated storage of flammable or volatile materials in general laboratory work areas.
  • Microprocessor temperature controller
  • Digital temperature display
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (⅜")
  • CFC-free refrigerant
No electrical components exist in the interior of the units and the compressor’s electrical components are sealed in a vapor-proof enclosure to reinforce safety.
Unit includes a series of temperature alarms for close temperature monitoring. They are built with high-density foam insulation and shelves for product storage. Require periodic defrosting to prevent ice build-up. Flammable storage units run on CFC-free refrigerant for economic efficiency and environmental safety.
Certifications: UL/C-UL listed.
Units meet NFPA guidelines 45 and 70
Ordering information: Two-year parts and labor warranty.