ESD Conductive Chair Mats, Transforming Technologies
Supplier: Transforming Technologies
FM73648 FM7CHAIR FM72448 FM73X75 FM74X75
76489-710EA 3321.55 USD
76489-710 76489-708 76489-706 76489-704 76489-712
ESD Conductive Chair Mats, Transforming Technologies
Floor Mats Permanent Floor Mats
The perfect solution for grounding chairs or carts in an industrial or manufacturing environment.
  • Rigid conductive vinyl
  • Black
  • 0.090" thick
  • 46×50" Chair mat available
The VinylSTAT FM7 is made of a rigid conductive vinyl that protects carpets from wear and tear while providing a path-to-ground for static charges. The ESD properties of the matting are permanent and are unaffected by damage caused by moving furniture. The material is also an excellent floor runner for general uses.
The FM7 mats are made from a single layer of conductive vinyl, die cut with a lip to fit under typical desks. Runners in various lengths and full 75′ long rolls are also available. The surface resistivity of the matting is 103 to 106.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and the recommendations of ESD STM7.1.