AccessPRO Turnstile with PGT120.COM, Transforming Technologies
Supplier: Transforming Technologies
PGT120K100
76490-160EA 19864.86 USD
76490-160
AccessPRO Turnstile with PGT120.COM, Transforming Technologies
Static Control Monitors
The PGT120 personnel grounding tester takes quick and accurate resistance measurements of the ESD grounding products and ensures they are working properly.
  • Independently test footwear and wrist strap grounding products
  • Pass/fail results are indicated audibly and visually
  • Mount via pedestal stand, wall mount, or in a turnstile
The unit operates with three independent measuring circuits for the left shoe, the right shoe, and the wrist strap. This makes it possible to measure all the values at the same time. The shoes can even be measured in a hands-free mode that allows staff members to be tested automatically when standing on the plate. It can also be connected to an automatic door opening system which allows entry with only passing results.