1,2-Diamino-9,10-anthraquinone ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 1,2-Diaminoanthraquinone

15220
76482-332EA 114.94 USD
76482-332
1,2-Diamino-9,10-anthraquinone ≥95%
1,2-Diamino-9,10-anthraquinone

The role of nitric oxide (NO) as a putative mediator of neuronal death can be understood best if NO is detected directly.

Formula: C₁₄H₁₀N₂O₂
MDL Number: MFCD00001219
CAS Number: 1758-68-5
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR