Direct cDNA Sequencing Kit, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
SQK-DCS109
76487-162EA 599 USD
76487-162
Direct cDNA Sequencing Kit, Oxford Nanopore Technologies
Nucleic Acid Reagents Next Generation Sequencing Reagents
The Direct cDNA Sequencing Kit enables the identification and quantification of full-length transcripts, without PCR bias, using nanopore sequencing.
  • Identify and quantify full-length transcripts
  • Explore differential gene expression
  • Characterise and quantify isoforms, splice variants, and fusion transcripts
  • Avoid bias and artefacts introduced through PCR
  • Multiplex ≤24 samples using Native Barcoding Expansion Packs
The Direct cDNA Sequencing Kit is used to prepare cDNA for nanopore sequencing without using PCR. The kit recommends an input of 100 ng poly-A+RNA.
Taking full-length poly-A+ RNA, complementary strand synthesis and strand switching are performed using kit-supplied oligonucleotides. The RNA template is then digested, and the second (complementary) strand is synthesised. Sequencing adapters supplied in the Direct cDNA Sequencing Kit are then ligated onto the dscDNA.
The Native Barcoding Expansion packs 1-12 (76487-118) and 13-24 (76487-120) can be used to multiplex up to 24 different RNA samples into a single flow cell.
For sample multiplexing see Native Barcoding expansion packs.
This product includes a flow cell priming kit.
Delivery information: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.