Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Roadside Safety Kits

https://us.vwr.com/stibo/smallweb/std.lang.all/97/40/18989740.jpg

Stowing easily in trunks, roadside safety kits adequately prepare drivers for any emergency on the roadway. The brightly colored reflective items alert traffic of immobile vehicle location and are weighted to stay upright despite the rapid rate of passing automobiles. Equipment is durably constructed to withstand inclement weather conditions that typically cause maintenance requirements. Compliant with transportation requirements, the roadside safety kits promote safer environments when drivers need to complete repairs.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"2"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (94013-724)

Supplier:  National Marker
Description:   Durable plastic carrying case contains three 41.91 cm (16.5") reflective triangles with weighted base and wire bracing.

Supplier:  National Marker
Description:   Heavy-duty steel carrying case contains three 41.91cm (16.5"), one 1.25kg (2.75lb) refillable portable fire extinguisher, three 20 minute flares with ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 2  of 2
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping