You Searched For: Soldering Stations
Unlike plug-in irons, soldering stations offer more control with the adjusting tip temperatures during fusion applications. The tip sensors automatically control heat levels for adequate protection when dealing with sensitive components and the provided sponges remove corrosion from ends for a strong working longevity. Since resting a hot iron on any surface can be extremely dangerous, the complete soldering station systems provide a safe support to secure the instruments without accidents.
Unlike plug-in irons, soldering stations offer more control with the adjusting tip temperatures during fusion applications. The tip sensors automatically control heat levels for adequate protection when dealing with sensitive components and the provided sponges remove corrosion from ends for a strong working longevity. Since resting a hot iron on any surface can be extremely dangerous, the complete soldering station systems provide a safe support to secure the instruments without accidents.
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (37697-504)
Supplier: Cooper Tools
Description: Contact pyrometer with large 20-gauge wire for use with WA2000 soldering iron analyzer.
Catalog Number: (76249-356)
Supplier: Cooper Tools
Description: This WX soldering station with an elegant design and solid improvements in performance offers the perfect combination of features to help the operatio...
Catalog Number: (76252-738)
Supplier: AMERICAN HAKKO PRODUCTS INC TE
Description: A complete, economical SMD hot air rework system, including the HAKKO FR-811 hot air station, bottom heater, handpiece rework fixture, board holder an...
Catalog Number: (76252-244)
Supplier: AMERICAN HAKKO PRODUCTS INC TE
Description: An advanced SMD rework station with vacuum pickup and a multitude of features for every need. When integrated with a bottom heater, board fixture and ...
Catalog Number: (76248-054)
Supplier: AMERICAN HAKKO PRODUCTS INC TE
Description: Self-contained desoldering and soldering station with a built-in vacuum pump. This versatile station uses the N3 series quick-change composite nozzles...
Catalog Number: (76244-432)
Supplier: TESTEQUITY LLC TE
Description: Soldering Aid
Catalog Number: (76253-714)
Supplier: OK INTERNATIONAL TE
Description: Metcal soldering stations with SmartHeat® technology. SmartHeat senses the thermal load and instantly delivers Power on Demand directly to the joint.
Catalog Number: (76253-322)
Supplier: OK INTERNATIONAL TE
Description: For use with Metcal™ precision tweezer hand-piece on MFR series systems.
Supplier: OK INTERNATIONAL TE
Description: Coil assembly for MFR-HDS and MFR-H5-DS Desoldering Hand-pieces.
Catalog Number: (76248-382)
Supplier: JBC TOOLS USA INC. TE
Description: These control units manages for soldering or desoldering.
Catalog Number: (300063-152)
Supplier: Safety & Industrial Supplies
Description: SOLDERING STATION 50W 120 V ET
Catalog Number: (76248-702)
Supplier: JBC TOOLS USA INC. TE
Description: The DME manages up to 4 tools simultaneously and it is fully compatible with most of tools.
Supplier: Pace
Description: Stations offer high capacity, low temperature SMT/thru-hole soldering and desoldering
UL Listed
Catalog Number: (76251-120)
Supplier: AMERICAN HAKKO PRODUCTS INC TE
Description: FM-205 is a “shop-air” desoldering and soldering station. The shop-air feature provides for higher vacuum pressure and increased suction flow in compa...
Catalog Number: (76268-272)
Supplier: JBC TOOLS USA INC. TE
Description: The best solution for reworking very small-sized components requiring the highest precision such as chips 0402, 0201, 01005.
Supplier: AMERICAN HAKKO PRODUCTS INC TE
Description: These stations are designed with vacuum pick up function for easy and safe removal of components.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 17
no targeter for Bottom