Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

You Searched For:

Soldering Irons

32  results were found
https://us.vwr.com/stibo/smallweb/std.lang.all/98/08/18989808.jpg

Soldering irons fuse equipment pieces together that would normally be difficult to combine. Designed for maximum heat transfer from electricity or butane fuel sources to the tip, the devices typically run at a fixed temperature for optimal control. The handheld tools accept interchangeable tips in various styles and sizes to fit specific applications and working components. With a lightweight and ergonomic handle, the soldering irons will remain comfortable even during extended use.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"32"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  JBC TOOLS USA INC. TE
Description:   These soldering iron handle for general purposes is suitable for both SMD components and for jobs with high power requirements.
Catalog Number: (76243-534)

Supplier:  JBC TOOLS USA INC. TE
Description:   The smallest handle for soldering very small-sized components requiring the greatest precision.
Supplier:  THERMALTRONICS USA INC TE
Description:   ESD Solder Handle W/RMP-1 for Soldering Station.

Supplier:  JBC TOOLS USA INC. TE
Description:   This soldering iron handle is perfect for intensive soldering jobs requiring continued high thermal power such as large ground planes, production of s...
Supplier:  Safety & Industrial Supplies
Description:   Use to braze with special alloy welding rods, heat and bend thin metals, loosen rusted bolts, pop dents out of metal, shrink and terminate heavy wall ...
Supplier:  Cooper Tools
Description:   Modular soldering irons feature a heater for thread-on tips, a 1.5m (5') three-wire grounded cord, and a handle with acid- and flux-resistant cushione...
UL Listed cUL Listed CSA Certified
Supplier:  AMERICAN HAKKO PRODUCTS INC TE
Description:   Micro Soldering Iron Conversion Kit used with FM-203 Dual Port Soldering System.
Catalog Number: (470123-386)

Supplier:  JA ASSOCIATES
Description:   30 Watt Soldering Iron
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 32
  1  2  3  4  Next