    Sort by:
  • Search Within Results
Desoldering Braids
https://us.vwr.com/stibo/smallweb/std.lang.all/51/08/18945108.jpg
After placing over the joint and pressing down with an iron, desoldering braids capture the filling metal using capillary action. Removing the weaved coil while the solder is still molten, the filled portion of the roll can simply be cut and discarded. The wicking action of the twisted copper wire protects equipment against heat damage, reduces cleanup time, and offers a higher filling capacity. Desoldering braids provide residue-free cleaning and corrective fixes should soldering mistakes occur.
SearchPresentationType-VERTICAL
SearchResultCount:"5"
  List View Easy View
Sort by:
Supplier:  ITW Chemtronics
Description:   Desoldering braids significantly reduce rework/repair time and minimize the risk of heat damage
Supplier:  ITW Chemtronics
Description:   Soldering masks protect PC boards, plated thru-holes, contacts, pins, posts, terminals, and gold fingers during wave soldering
Supplier:  ITW Chemtronics
Description:   Ultrapure, oxygen-free copper braids which quickly and completely remove solder from circuit boards and components.
Supplier:  ITW Chemtronics
Description:   These ultrapure, oxygen-free copper braids quickly and completely remove solder from circuit boards and components
Supplier:  ITW Chemtronics
Description:   Thixotropic, synthetic acrylic latex masking compounds to temporarily mask areas of printed circuit boards
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 5  of 5