Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Apoptosis Inducers

https://us.vwr.com/stibo/smallweb/std.lang.all/49/05/18944905.jpg

Apoptosis inducers initiate stimuli in a regulated process that cause intracellular changes and inevitably death. Used with multicellular organism cells, the high quality solutions have either mitochondrial intrinsic path or extrinsic path activation mechanisms based on the sample sensitivity. The programmed death generators may target specific processes to prevent the growth of malignant cells. Possessing anticancer or antitumor properties, apoptosis inducers are essential medical researching agents for improving and creating advanced treatment options.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"52"
Sort by:
 
 
 
 

Supplier:  MilliporeSigma
Description:   Most commonly-used phorbol ester
Catalog Number: (80052-248)
Supplier:  MilliporeSigma
Description:   A major aldehydic product formed by peroxidation of [omega]-6-unsaturated fatty acids
Catalog Number: (80055-474)
Supplier:  MilliporeSigma
Description:   A cell-permeable tumor-promoting sesquiterpene lactone that releases Ca2+ by inhibiting endoplasmic reticular Ca2+-ATPase (IC50=4–13nM)
SDS
Supplier:  MilliporeSigma
Description:   Has immunomodulatory properties in vitro and in vivo
Supplier:  BioVision
Description:   MDV-3100 (Enzalutamide) is an androgen-receptor (AR) antagonist (IC₅₀ = 36 nM).
SDS
Supplier:  MilliporeSigma
Description:   A utility carrier of nitric oxide. Breaks down to produce nitric oxide and glutathione at pH 7.4. Causes smooth muscle relaxation and inhibits platelet aggreration. Purity: >=95% by HPLC. Soluble in water. CAS: 57564-91-7. FW: 336.3. Light pink solid, 10mg.
Supplier:  MilliporeSigma
Description:   Stimulates the activity of inducible nitric oxide synthase
Supplier:  MilliporeSigma
Description:   Anticancer, antiviral agent that is especially effective against leukemias.
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 52
  1  2  3  4  5  6  7  Next