You Searched For: Heel and Toe Grounders
Constructed from durable, non-slip materials, the heel grounders provide protection for staff on the move. These heel grounders connect the person wearing them to the ground via a proper floor mat or flooring material. When worn on both feet, the devices provide consistent grounding even while in motion. Toe-style grounders feature a conductive cup that fits securely on the toe of footwear and is held in place by an elastic ankle strap.
Constructed from durable, non-slip materials, the heel grounders provide protection for staff on the move. These heel grounders connect the person wearing them to the ground via a proper floor mat or flooring material. When worn on both feet, the devices provide consistent grounding even while in motion. Toe-style grounders feature a conductive cup that fits securely on the toe of footwear and is held in place by an elastic ankle strap.
35  results were found
SearchResultCount:"35"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (76238-052)
Supplier: Desco
Description: The Shoe Grounder Assembly is designed to effectively ground mobile personnel when they are in contact with conductive and dissipative flooring.
Catalog Number: (14224-330)
Supplier: Transforming Technologies
Description: The sport cup heel grounder from Transforming Technologies is ideal for boots and cross-training athletic shoes
Supplier: Transforming Technologies
Description: Full coverage sole grounders provide 20 times the surface contact of heel grounders.
Supplier: Desco
Description: Premium Foot Grounder, Small
Catalog Number: (26902-604)
Supplier: Desco
Description: This grounder incorporates a three-layer yellow heel cup that will not mar shoes or floors
Product available on GSA Advantage®
Supplier: Desco
Description: Adjustable heel foot grounder with non-marring interior layer.
Supplier: Desco
Description: Sole Foot Grounders provide a large contact point on the ball of the foot, which forms a more consistent path to ground.
Product available on GSA Advantage®
Catalog Number: (76474-662)
Supplier: Transforming Technologies
Description: Ground mobile personnel with large size footwear with long-lasting stretch Hook and Loop ESD heel grounders.
Catalog Number: (76489-732)
Supplier: Transforming Technologies
Description: Disposable heel grounders are comfortable to wear, easily identifiable, and use the entire volume of the fabric to provide protection.
Catalog Number: (14223-828)
Supplier: Statico
Description: The S1100 Series is an easy to use disposable heel grounder that dissipates the static charges effectively, while offering convenience of use
Catalog Number: (76221-338)
Supplier: High-Tech Conversions
Description: These neon yellow ESD heel straps provide a visual aid to ensure all personnel are in compliance within the lab.
Supplier: Electron Microscopy Sciences
Description: low cost premium grade non-marking conductive rubber, highly abrasion and wear resistant, adjustable Velcro fastening system, Includes:18” conductive tab and 1½”, two-layer, rubber cup, built in resistor, charge
Minority or Woman-Owned Business Enterprise
Supplier: Transforming Technologies
Description: Heel Grounders are constructed from durable materials and feature a stretch hook and loop closure strap to maximize fit and comfort.
Catalog Number: (76209-916)
Supplier: Transforming Technologies
Description: Disposable Heel Grounder is the perfect solution for plant visitors and other temporary personnel who enter static controlled environments with ESD pr...
Catalog Number: (76243-212)
Supplier: Transforming Technologies
Description: This heel grounder is constructed from durable materials and features a classic hi-visibility yellow/green hook and loop closure with an elastic inser...
Supplier: Transforming Technologies
Description: Full coverage sole grounders provide 20 times the surface contact of heel grounders, with a wider and more consistent contact area to ensure grounding...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 35
no targeter for Bottom