VWR® Standard Series Convertible Freezer/Refrigerator
Supplier: VWR, part of Avantor
VWR®
VWV20RPSA VWV20FPSA
76470-402EA 2544 USD
76470-402 76449-108
VWR® Standard Series Convertible Freezer/Refrigerator
Refrigerators
VWR® Standard Series convertible freezers and refrigerators offer economical and simple sample storage while also offering increased flexiblity of operating as either a refrigerator or a freezer with the push of a button.
  • The VWV20FPSA ships as an auto defrost freezer and the VWV20RPSA ships as a refrigerator but both easily converts to a refrigerator or an auto defrost freezer respectively
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish