Valve Supplies for HPLC System, Agilent Technologies
Supplier: Agilent Technologies
0100-2195 0101-0921 0101-1390 0101-1050 5068-0006 0101-1416 0101-1409 0101-1417 5068-0007 0101-1422
101472-232EA 1129.12 USD
101472-232 101472-088 101471-420 101472-236 101471-600 101479-528 101449-222 101479-766 101479-762 101472-230
Valve Supplies for HPLC System, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Vespel is a polyimide with low wear and high chemical resistance. Vespel tolerates a pH range of 0 to 10.
PEEK offers a high chemical resistance and flexibility and will tolerate the entire pH range from 0 to 14.
Tefzel is ideal for use in applications where PEEK cannot be used, such as methylene chloride or DMSO in higher concentrations.