Alphagen® EtBr Destroyer Bags and Spray, Bulldog Bio
Supplier: Bulldog Bio
Alphagen®
AEDB10 AEDB20 AEDS1000 AEDM521 AEDS2200 AEDB50 AEDS2210
103959-142PK 166.15 USD
103959-142 103959-130 103959-140 103959-134 103959-132 103959-138 103959-136
Alphagen® EtBr Destroyer Bags and Spray, Bulldog Bio
Bags Waste Disposal Bags
EtBr Destroyer efficiently cleans up ethidium bromide (EtBr), resulting in the safe disposal of biproducts.
  • Neutralize toxic ethidium bromide
  • Effective SYBR Green, SYBR Gold, and SYPRO Orange
  • Store at room temperature
EtBr Destroyer is formulated to efficiently break down and clean up ethidium bromide (EtBr), resulting in only non-fluorescent, non-mutagenic biproducts. Available in two convenient formats for the cleaning and treatment of both solid and liquid EtBr contaminants, it has also proven effective against SYBR Dyes. The ready-pack EtBr Destroyer destaining bags are ideal for the treatment of liquid solutions or agarose gels that contain EtBr. For contact contamination issues, the EtBr Destroyer Spray can be used for the treatment of solid materials that have been exposed to EtBr including electrophoresis gel boxes, glassware, paper towels, gloves, laboratory equipment, bench surfaces, and much more.