Digital Water Bath
Supplier: Edvotek
539
470334-072EA 539 USD
470334-072
Digital Water Bath
Baths Water Baths, Unstirred
This classic Edvotek® water bath has been updated and improved - 60% larger than its predecessor and now has a built-in timer. The water bath still includes a digital temperature control and low-water sensor to prevent burn-outs.
The stainless steel chamber is corrosion resistant and temperature controlled from ambient to 99 °C while using the included cover.
Delivery information: Cover included.