Masterflex® Remote Control Connector, Cole-Parmer
Supplier: Cole Parmer
07595-45
BR7595-45EA 104.69 USD
BR7595-45
Masterflex® Remote Control Connector, Cole-Parmer
Connectors Connectors, Tubing
Create your own cable for optimization of a Masterflex® drive.
  • Choose the correct connection for a Masterflex® pump
Combine these connectors with 20 to 22 gauge shielded wire to create an interface cable. Maximum cable length is 1000' (300 m).