Masterflex® Dispensing Tips, Polypropylene, Cole-Parmer
Supplier: Cole Parmer
7822662
97057-296EA 126.18 USD
97057-296
Masterflex® Dispensing Tips, Polypropylene, Cole-Parmer
Fittings and Connectors
Attach with just a twist.
  • Constructed of durable polypropylene
  • Easily attach flexible tubing to the hose barb connection
These luer fittings are ideal for a variety of critical applications, including connection to syringes or small diameter flexible tubing.