Tris(pentafluorophenyl)borane 97%

Supplier: Ambeed

Warning

Synonyms: Tris(perfluorophenyl)borane

A118336-5G A118336-1G
103830-404EA 102 USD
103830-404 103799-036
Tris(pentafluorophenyl)borane 97%
Tris(pentafluorophenyl)borane

Caution: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₁₈BF₁₅
MW: 511.98 g/mol
Boiling Pt: 327.3 °C (760 mm Hg)
Melting Pt: 126…131 °C
Storage Temperature: Freezer
MDL Number: MFCD00269813
CAS Number: 1109-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR