Tricyclohexylphosphonium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A239729-5G A239729-10G A239729-5G A239729-500G A239729-25G A239729-100G
76786-508EA 345.05 USD
76786-508 77090-244 103851-102 76786-506 77090-246 76786-504
Tricyclohexylphosphonium tetrafluoroborate 97%
Tricyclohexylphosphonium tetrafluoroborate

Caution: for Research Use Only

Formula: C₁₈H₃₄BF₄P
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD03840580
CAS Number: 58656-04-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR