Thianthrene 98%

Supplier: Ambeed
Warning

A272407-5G A272407-5G A272407-100G A272407-25G
77305-110EA 6 USD
77305-110 76723-062 103853-006 77305-108
Thianthrene 98%
Thianthrene

Caution: for Research Use Only

Formula: C₁₂H₈S₂
MW: 216.33 g/mol
Boiling Pt: 344.9 °C (760 mm Hg)
Melting Pt: 151…155 °C
Flash Pt: 170.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00005065
CAS Number: 92-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR