(±)-1-Boc-3-hydroxypyrrolidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-hydroxypyrrolidine-1-carboxylate
, 3-Hydroxypyrrolidine, N-BOC protected
A213455-10G
A213455-5G
A213455-5G
A213455-100G
A213455-25G
A213455-500G
77087-524EA
8.24
USD
77087-524
103849-008
77261-132
77087-522
(±)-1-Boc-3-hydroxypyrrolidine 98%
(±)-1-Boc-3-hydroxypyrrolidine
Caution: for Research Use Only
Formula:
C₉H₁₇NO₃ MW: 187.24 g/mol Boiling Pt: 273.3±33.0 °C (760 mm Hg) Melting Pt: 57…63 °C Flash Pt: 119.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038535 CAS Number: 103057-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...