Sodium tetrachloroaurate dihydrate 98% Au > 48%

Supplier: Ambeed
Warning

Synonyms: Sodium tetrachloroaurate(III) dihydrate

A129421-250MG A129421-100MG A129421-5G A129421-1G
76718-898EA 24.72 USD
76718-898 103801-282 76718-900 103832-166
Sodium tetrachloroaurate dihydrate 98% Au > 48%
Sodium tetrachloroaurate dihydrate

Caution: for Research Use Only

Formula: NaAuCl₄·2H₂O
MW: 397.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149162
CAS Number: 13874-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR