6-Quinoxalinecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Quinoxaline-6-carboxylic acid
A152455-1G
A152455-5G
103842-562EA
49
USD
103842-562
103804-394
6-Quinoxalinecarboxylic acid 97%
6-Quinoxalinecarboxylic acid
Caution: Sealed in dry, at room temperature.
Formula:
C₉H₆N₂O₂ MW: 174.16 g/mol Boiling Pt: 355.1 °C (760 mm Hg) Melting Pt: 250 °C Flash Pt: 168.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD01365834 CAS Number: 6925-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...