Pyridazine-3-carboxylic acid 97%

Warning

A339951-1G A339951-5G
103836-776EA 94.62 USD
103836-776 103825-802
Pyridazine-3-carboxylic acid 97%
Pyridazine-3-carboxylic acid

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Boiling Pt: 404.2±18.0 °C (760 mm Hg)
Melting Pt: 199…201 °C
Flash Pt: 198.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01646333
CAS Number: 2164-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR