1-Pyreneboronic acid 97%

Supplier: Ambeed

Warning

Synonyms: Pyren-1-ylboronic acidPyrene-1-boronic acid

A109255-1G A109255-5G
103797-476EA 9 USD
103797-476 103823-270
1-Pyreneboronic acid 97%
1-Pyreneboronic acid

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₆H₁₁BO₂
MW: 246.07 g/mol
Boiling Pt: 509.4 °C (760 mm Hg)
Melting Pt: 247…251 °C
Flash Pt: 261.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04974062
CAS Number: 164461-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR