Potassium-3-hydroxyphenyltrifluoroborate 97%

Warning

Synonyms: Potassium trifluoro(3-hydroxyphenyl)borate

A279574-1G A279574-5G
103853-418EA 42.69 USD
103853-418 103813-472
Potassium-3-hydroxyphenyltrifluoroborate 97%
Potassium 3-hydroxyphenyltrifluoroborate

Caution: Inert atmosphere, room temperature

Formula: C₆H₅BF₃KO
MW: 200.0100 g/mol
Melting Pt: 170…190 °C
Storage Temperature: Ambient
MDL Number: MFCD07784422
CAS Number: 871231-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR