3-Oxetanone 98%

Supplier: Ambeed

Danger

Synonyms: Oxetan-3-one

A383679-1G A383679-5G
103837-782EA 18 USD
103837-782 103826-744
3-Oxetanone 98%
3-Oxetanone

Caution: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 140 °C (760 mm Hg)
Flash Pt: 53 °C
Storage Temperature: Freezer
MDL Number: MFCD09263255
CAS Number: 6704-31-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR