5-Oxazolecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Oxazole-5-carboxylic acid

A179635-10G A179635-1G A179635-5G
77011-950EA 70.04 USD
77011-950 103845-802 103807-450
5-Oxazolecarboxylic acid 98%
5-Oxazolecarboxylic acid

Caution: for Research Use Only

Formula: C₄H₃NO₃
MW: 113.07 g/mol
Boiling Pt: 289.3 °C (760 mm Hg)
Melting Pt: 195…197 °C
Flash Pt: 128.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04114931
CAS Number: 118994-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR