N-Acetylguanidine 97%

Supplier: Ambeed

Warning

Synonyms: N-Carbamimidoylacetamide1-Acetylguanidine

A370145-5G
103837-470EA 13 USD
103837-470
N-Acetylguanidine 97%
N-Acetylguanidine

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₃H₇N₃O
MW: 101.11 g/mol
Melting Pt: 181…183 °C
Flash Pt: 98.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00075615
CAS Number: 5699-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR