1-Boc-4-bromopiperidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-tert-Butoxycarbonyl-4-bromopiperidine
, 1-Boc-4-bromopiperidine
, tert-Butyl 4-Bromopiperidine-1-carboxylate
, N-Boc-4-Bromopiperidine
, 4-Bromopiperidine-1-carboxylic acid tert-Butyl ester
A279305-1G
A279305-5G
A279305-10G
A279305-1G
A279305-100G
A279305-250MG
A279305-25G
A279305-5G
77014-578EA
5.15
USD
77014-578
103813-462
77014-576
77093-344
77014-574
77305-326
77305-324
103853-406
1-Boc-4-bromopiperidine 98%
1-Boc-4-bromopiperidine
Caution: for Research Use Only
Formula:
C₁₀H₁₈BrNO₂ MW: 264.1594 g/mol Boiling Pt: 296.6 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115039 CAS Number: 180695-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...