1,4-Naphthalenedicarboxylic acid 98%

Supplier: Ambeed

Warning

Synonyms: Naphthalene-1,4-dicarboxylic acid

A419708-5G
103838-420EA 8 USD
103838-420
1,4-Naphthalenedicarboxylic acid 98%
1,4-Naphthalenedicarboxylic acid

Caution: Sealed in dry, at room temperature.

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Boiling Pt: 490.2 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 264.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00014312
CAS Number: 605-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR