Methyl-5-hydroxypicolinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-hydroxypyridine-2-carboxylate
A114747-1G
A114747-250MG
A114747-1G
A114747-10G
A114747-100G
A114747-5G
A114747-5G
76766-078EA
714.82
USD
76766-078
103824-306
103798-464
77311-130
76766-082
77311-128
76766-080
Methyl-5-hydroxypicolinate 97%
Methyl 5-hydroxypicolinate
Caution: for Research Use Only
Formula:
C₇H₇NO₃ MW: 153.1354 g/mol Melting Pt: 193.5…195.5 °C Flash Pt: 180.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD05256426 CAS Number: 30766-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...