Methyl-4-methoxyacetoacetate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyacetoacetic acid methyll esterMethyl 4-methoxy-3-oxobutanoate

A314166-5G A314166-500G
76648-188EA 161.71 USD
76648-188 103835-946
Methyl-4-methoxyacetoacetate 95%
Methyl 4-methoxyacetoacetate

Caution: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.1412 g/mol
Melting Pt: 303 °C
Flash Pt: 89.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010183
CAS Number: 41051-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR