Methyl-4-ethynylbenzoate 98%

Supplier: Ambeed

Warning

Synonyms: Methyl 4-ethynylbenzoate

A245615-5G A245615-1G
103811-658EA 20 USD
103811-658 103851-490
Methyl-4-ethynylbenzoate 98%
Methyl 4-ethynylbenzoate

Caution: Inert atmosphere, 2…8 °C.

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 239.4 °C (760 mm Hg)
Melting Pt: 88…95 °C
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168820
CAS Number: 3034-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR