Methyl-4-(hydroxymethyl)picolinate 97%

Warning

Synonyms: Methyl 4-(hydroxymethyl)picolinateMethyl 4-(hydroxymethyl)pyridine-2-carboxylate

A108076-1G A108076-5G A108076-250MG
103823-064EA 246.92 USD
103823-064 103797-290
Methyl-4-(hydroxymethyl)picolinate 97%
Methyl 4-(hydroxymethyl)picolinate

Caution: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834350
CAS Number: 317335-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR