Methyl-3-hydroxyisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-hydroxyisonicotinate

A186912-10G A186912-1G A186912-5G A186912-250MG A186912-5G
76721-040EA 190.55 USD
76721-040 103846-566 77302-058 76721-042 103815-978
Methyl-3-hydroxyisonicotinate 98%
Methyl 3-hydroxyisonicotinate

Caution: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.1400 g/mol
Boiling Pt: 313.6 °C (760 mm Hg)
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00661299
CAS Number: 10128-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR