Methyl-3-bromo-4-methylbenzoate 98%

Supplier: Ambeed

Warning

Synonyms: Methyl 3-bromo-4-methylbenzoate

A103989-5G A103989-1G
103822-340EA 9 USD
103822-340 103796-628
Methyl-3-bromo-4-methylbenzoate 98%
Methyl 3-bromo-4-methylbenzoate

Caution: Inert atmosphere, room temperature

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 266.9 °C (760 mm Hg)
Melting Pt: 38…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00144769
CAS Number: 104901-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR