Methyl-2-methyl-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-5-nitrobenzoate

A212253-1G A212253-1G A212253-5G A212253-100G A212253-5G A212253-500G A212253-10G A212253-25G
77087-338EA 28.84 USD
77087-338 103818-180 77275-220 103848-880 77087-340 77275-218 77275-216 77303-238
Methyl-2-methyl-5-nitrobenzoate 97%
Methyl 2-methyl-5-nitrobenzoate

Caution: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Boiling Pt: 305.4 °C (760 mm Hg)
Flash Pt: 141.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00230865
CAS Number: 77324-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR