Methyl-2-fluoroisonicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-fluoroisonicotinate
A179912-5G
A179912-1G
103845-844EA
407
USD
103845-844
103807-490
Methyl-2-fluoroisonicotinate 98%
Methyl 2-fluoroisonicotinate
Caution: Inert atmosphere, 2…8 °C.
Formula:
C₇H₆FNO₂ MW: 155.1300 g/mol Boiling Pt: 227.7 °C (760 mm Hg) Flash Pt: 93.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11656063 CAS Number: 455-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...