Isoxazole-4-boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isoxazol-4-ylboronic acid
A258383-1G
A258383-250MG
A258383-5G
A258383-1G
76786-916EA
32.96
USD
76786-916
103852-244
77091-772
103812-370
Isoxazole-4-boronic acid 98%
Isoxazole-4-boronic acid
Caution: for Research Use Only
Formula:
C₃H₄BNO₃ MW: 112.8798 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06657892 CAS Number: 1008139-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...