4-Aminoisoxazole 97%

Supplier: Ambeed

Warning

Synonyms: Isoxazol-4-amine

A303650-1G A303650-5G
103814-386EA 208 USD
103814-386 103835-552
4-Aminoisoxazole 97%
4-Aminoisoxazole

Caution: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₃H₄N₂O
MW: 84.0767 g/mol
Boiling Pt: 234.7 °C (760 mm Hg)
Flash Pt: 95.7 °C
Storage Temperature: Freezer
MDL Number: MFCD08234629
CAS Number: 108511-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR