Imidazo[1,2-b]pyridazine-6-carboxylic acid 97%

Supplier: Ambeed

Warning

A167442-1G
103806-188EA 399 USD
103806-188
Imidazo[1,2-b]pyridazine-6-carboxylic acid 97%
Imidazo[1,2-b]pyridazine-6-carboxylic acid

Caution: Sealed in dry, at 2…8°C

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044772
CAS Number: 769109-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR