Europium(III) trifluoromethanesulfonate 98%

Supplier: Ambeed

Warning

Synonyms: Europium triflateEuropium(III) trifluoromethanesulfonate

A641946-5G
103840-344EA 76 USD
103840-344
Europium(III) trifluoromethanesulfonate 98%
Europium (III) trifluoromethanesulphonate

Caution: Inert atmosphere, 2…8 °C.

Formula: C₃EuF₉O₉S₃
MW: 599.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00209587
CAS Number: 52093-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR