Ethyl-5-bromothiophene-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-bromothiophene-2-carboxylate
A172317-10G
A172317-1G
A172317-1G
A172317-5G
A172317-5G
A172317-25G
103845-000EA
17.31
USD
103845-000
77301-194
76784-292
77082-198
103806-716
77082-196
Ethyl-5-bromothiophene-2-carboxylate 98%
Ethyl 5-bromothiophene-2-carboxylate
Caution: for Research Use Only
Formula:
C₇H₇BrO₂S MW: 235.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683089 CAS Number: 5751-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...