Ethyl-5-bromopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-bromopicolinate

A101561-250MG A101561-1G A101561-5G A101561-1G A101561-5G
103821-712EA 117.69 USD
103821-712 77107-206 103796-026
Ethyl-5-bromopicolinate 98%
Ethyl 5-bromopicolinate

Caution: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Boiling Pt: 303.7 °C (760 mm Hg)
Melting Pt: 63…64 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00234415
CAS Number: 77199-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR