Ethyl-5-amino-1-benzofuran-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 5-Aminobenzofuran-2-carboxylic acid ethyl esterEthyl 5-aminobenzo[b]furan-2-carboxylateEthyl 5-Aminobenzofuran-2-carboxylate

A221538-10G A221538-5G A221538-250MG A221538-5G A221538-1G A221538-1G
76800-886EA 5.15 USD
76800-886 76800-884 103818-932 103849-812 77088-408 77088-410
Ethyl-5-amino-1-benzofuran-2-carboxylate 95%
Ethyl-5-amino-1-benzofuran-2-carboxylate

Caution: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Boiling Pt: 348.1 °C (760 mm Hg)
Melting Pt: 54…56 °C
Flash Pt: 164.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275091
CAS Number: 174775-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR