Ethyl trimethylacetopyruvate 98%

Supplier: Ambeed

Warning

Synonyms: Ethyl 5,5-dimethyl-2,4-dioxohexanoate

A158792-5G A158792-1G
103843-386EA 48 USD
103843-386 103805-172
Ethyl trimethylacetopyruvate 98%
Ethyl trimethylacetopyruvate

Caution: Sealed in dry, at 2…8°C

Formula: C₁₀H₁₆O₄
MW: 200.24
Boiling Pt: 68 °C (0.5 mm Hg)
Flash Pt: 106.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052319
CAS Number: 13395-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR